Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Diiodosalicylic acid lithium salt hydrate, 95%, Thermo Scientific™

Catalog No. AAA1578406
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1578406 5 g
AAA1578414 25 g
AAA1578422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1578406 Supplier Thermo Scientific Chemicals Supplier No. A1578406

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 653-14-5
Molecular Formula C7H3I2LiO3
Molecular Weight (g/mol) 395.85
MDL Number MFCD00156142,MFCD00002443
InChI Key HLBRJWWTLIAOTE-UHFFFAOYSA-M
Synonym lithium 3,5-diiodosalicylate, lithium 2-hydroxy-3,5-diiodobenzoate, benzoic acid, 2-hydroxy-3,5-diiodo-, monolithium salt, 3,5-diiodosalicylic acid lithium salt, 2-hydroxy-3,5-diiodobenzoic acid lithium salt, 3,5-diiodo-2-hydroxybenzoic acid lithium salt, benzoic acid, 2-hydroxy-3,5-diiodo-, lithium salt 1:1, 3,5-diiodosalicylic acid lithium salt hydrate, acmc-1ark0, 2-hydroxy-3,5-diiodobenzoic acid, lithium salt
PubChem CID 2735070
SMILES [Li+].OC1=C(I)C=C(I)C=C1C([O-])=O

Specifications

Quantity 5 g
IUPAC Name lithium;2-hydroxy-3,5-diiodobenzoate
Formula Weight 413.86 (395.85 Anhydrous)
Percent Purity 95%
Chemical Name or Material 3,5-Diiodosalicylic acid lithium salt hydrate
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
TSCA TSCA
Recommended Storage −30° to −10°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.