Learn More
Selleck Chemical LLC (-)-Borneol S3977-25mg
SDP
Supplier:  Selleck Chemical LLC S397725MG
Encompass_Preferred
(-)-Borneol (L-Borneol) one of enantiomers of borneol has a camphoraceous and evident woody odor (-)-Borneol has a highly efficacious positive modulating action at GABA receptor with an EC50 of 237 M
Catalog No. 50-310-8394
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.