Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxybenzoyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D260925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D260925G 25 g
D26095G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D260925G Supplier TCI America Supplier No. D260925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17213-57-9
Molecular Formula C9H9ClO3
Molecular Weight (g/mol) 200.62
MDL Number MFCD00000676
InChI Key FTHPLWDYWAKYCY-UHFFFAOYSA-N
PubChem CID 87003
IUPAC Name 3,5-dimethoxybenzoyl chloride
SMILES COC1=CC(=CC(OC)=C1)C(Cl)=O

Specifications

Color White-Yellow
Quantity 25 g
UN Number 3261
Synonym benzoyl chloride, 3,5-dimethoxy, 3,5-dimethoxy benzoyl chloride, 3,5-dimethoxy-benzoyl chloride, acmc-1c560, benzoyl chloride,3,5-dimethoxy, 3,5-dimethoxybenzoic acid chloride, 3,5-dimethoxybenzoyl chloride, 3,5-dimethoxy-1-benzenecarbonyl chloride
Formula Weight 200.62
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Dimethoxybenzoyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.