Learn More
Medchemexpress LLC 3,5-dimethoxybenzyl alcohol | 705-76-0 | MFCD00004641 | 99.9% | 168.19 g·mol⁻¹ | C9H12O3 | 100 G

Supplier: Medchemexpress LLC HYW017093100G
3,5-dimethoxybenzyl alcohol is a phenolic benzyl alcohol used as a research reagent and intermediate in organic synthesis. It is reported from natural sources such as Pinus sylvestris and is supplied with supporting analytical documentation for laboratory use.
- High purity: 99.90% (HPLC).
- Chemical formula: C9H12O3.
- Molecular weight: 168.19 g·mol⁻¹.
- Storage: store at room temperature (stable 3 years); in solvent -80°C (2 years) or -20°C (1 year).
- Suitable as an intermediate in organic synthesis and research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.