Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxybenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. D259425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D259425G 25 g
D25945G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D259425G Supplier TCI America Supplier No. D259425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 705-76-0
Molecular Formula C9H12O3
Molecular Weight (g/mol) 168.19
MDL Number MFCD00004641
InChI Key AUDBREYGQOXIFT-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzyl alcohol, 3,5-dimethoxyphenyl methanol, 3,5-dimethoxybenzylalcohol, benzenemethanol, 3,5-dimethoxy, 3,5-dimethoxy benzyl alcohol, 3,5-dimethoxyphenyl methan-1-ol, 3,5-dimethoxybenzyl alcohole, pubchem7732, 3,5-dimethoxy-benzylalcohol, 3,5-dimethoxy benzylalcohol
PubChem CID 69718
IUPAC Name (3,5-dimethoxyphenyl)methanol
SMILES COC1=CC(OC)=CC(CO)=C1

Specifications

Melting Point 50°C
Color White
Boiling Point 171°C
Quantity 25 g
Formula Weight 168.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxybenzyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.