Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxybenzyl Bromide 97.0+%, TCI America™
SDP

Catalog No. D265725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D265725G 25 g
D26575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D265725G Supplier TCI America Supplier No. D265725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 877-88-3
Molecular Formula C9H11BrO2
Molecular Weight (g/mol) 231.089
MDL Number MFCD01321368
InChI Key BTHIGJGJAPYFSJ-UHFFFAOYSA-N
Synonym 3,5-dimethoxybenzyl bromide, 1-bromomethyl-3,5-dimethoxybenzene, 3,5-dimethoxybenzylbromide, benzene, 1-bromomethyl-3,5-dimethoxy, 3,5-dimethoxy benzyl bromide, alpha-bromo-3,5-dimethoxytoluene, 5-bromomethyl-1,3-dimethoxybenzene, 5-bromomethyl resorcinol dimethyl ether, pubchem13588, acmc-209qq1
PubChem CID 1274490
IUPAC Name 1-(bromomethyl)-3,5-dimethoxybenzene
SMILES COC1=CC(=CC(=C1)CBr)OC

Specifications

Melting Point 73°C
Color White-Yellow
Quantity 25 g
UN Number 3261
Formula Weight 231.09
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethoxybenzyl Bromide
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.