Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethoxytoluene 97.0+%, TCI America™
SDP

Catalog No. D252625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D252625G 25 g
D25265G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D252625G Supplier TCI America Supplier No. D252625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4179-19-5
Molecular Formula C9H12O2
Molecular Weight (g/mol) 152.19
MDL Number MFCD00015435
InChI Key RIZBLVRXRWHLFA-UHFFFAOYSA-N
Synonym 3,5-dimethoxytoluene, orcinol dimethyl ether, benzene, 1,3-dimethoxy-5-methyl, 5-methylresorcinol dimethyl ether, 3,5-dimethoxy toluene, toluene, 3,5-dimethoxy, 1,3-dimethoxy-5-methyl-benzene, pubchem17775, acmc-1ak5i
PubChem CID 77844
IUPAC Name 1,3-dimethoxy-5-methylbenzene
SMILES COC1=CC(OC)=CC(C)=C1

Specifications

Color Yellow
Boiling Point 112°C
Quantity 25 g
Formula Weight 152.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethoxytoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.