Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-1-phenyl-1H-pyrazole, 97%, Thermo Scientific™

Catalog No. AAB2548906
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2548906 5 g
AAB2548914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2548906 Supplier Thermo Scientific Chemicals Supplier No. B2548906

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1131-16-4
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD00020728
InChI Key ULPMPUPEFBDQQA-UHFFFAOYSA-N
PubChem CID 70800
IUPAC Name 3,5-dimethyl-1-phenylpyrazole
SMILES CC1=CC(=NN1C2=CC=CC=C2)C

Specifications

Boiling Point 144°C to 145°C (12 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 132902
Synonym 3,5-dimethyl-1-phenyl-1h-pyrazole, 1h-pyrazole, 3,5-dimethyl-1-phenyl, dimethylphenylpyrazole, unii-0sb52bxx7m, 1h-pyrazole,3,5-dimethyl-1-phenyl, 0sb52bxx7m, 1,3-dimethyl-5-phenylpyrazole, phenyldimethylpyrazole, enamine_001158, pyrazole, dimethylphenyl
Formula Weight 172.23
Percent Purity 97%
Chemical Name or Material 3,5-Dimethyl-1-phenyl-1H-pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.