Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-4-nitroisoxazole 98.0+%, TCI America™
SDP

Catalog No. D450625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D450625G 25 g
D45065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D450625G Supplier TCI America Supplier No. D450625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1123-49-5
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.11
MDL Number MFCD00051656
InChI Key PMQFLWLYAXYFHG-UHFFFAOYSA-N
PubChem CID 261410
IUPAC Name 3,5-dimethyl-4-nitro-1,2-oxazole
SMILES CC1=C(C(C)=NO1)[N+]([O-])=O

Specifications

Melting Point 63°C
Color White-Yellow
Quantity 25 g
Formula Weight 142.11
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-4-nitroisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.