Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethyl-4-nitropyrazole 98.0+%, TCI America™
SDP

Catalog No. D442525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D442525G 25 g
D44255G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D442525G Supplier TCI America Supplier No. D442525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14531-55-6
Molecular Formula C5H7N3O2
Molecular Weight (g/mol) 141.13
MDL Number MFCD00052513
InChI Key OFQCJVVJRNPSET-UHFFFAOYSA-N
PubChem CID 146172
IUPAC Name 3,5-dimethyl-4-nitro-1H-pyrazole
SMILES CC1=C(C(=NN1)C)[N+](=O)[O-]

Specifications

Melting Point 127°C
Color White-Yellow
Boiling Point 325°C
Quantity 25 g
Formula Weight 141.13
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dimethyl-4-nitropyrazole
RTECSNumber UQ6585000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.