Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylbenzaldehyde 96.0+%, TCI America™
SDP

Catalog No. D349825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D349825G 25 g
D34985G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D349825G Supplier TCI America Supplier No. D349825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5779-95-3
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00082777
InChI Key NBEFMISJJNGCIZ-UHFFFAOYSA-N
Synonym benzaldehyde, 3,5-dimethyl, 3,5-dimethyl benzaldehyde, m-xylene-5-carboxaldehyde, 3,5-dimethyl-benzaldehyde, pubchem15515, acmc-1arkp, 3,5-dimehtylbenzaldehyde, #, ksc494c0b, 3,5-dimethylbenzaldehyde
PubChem CID 34225
IUPAC Name 3,5-dimethylbenzaldehyde
SMILES CC1=CC(=CC(=C1)C=O)C

Specifications

Melting Point 9°C
Color Yellow
Boiling Point 108°C
Quantity 25 g
Formula Weight 134.18
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 3,5-Dimethylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.