Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylcyclohexanol (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. D070225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D070225ML 25 mL
D0702500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D070225ML Supplier TCI America Supplier No. D070225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5441-52-1
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.215
MDL Number MFCD00001447
InChI Key WIYNOPYNRFPWNB-UHFFFAOYSA-N
PubChem CID 21584
IUPAC Name 3,5-dimethylcyclohexan-1-ol
SMILES CC1CC(CC(C1)O)C

Specifications

Color White
Boiling Point 186°C
Quantity 25 mL
Formula Weight 128.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 3,5-Dimethylcyclohexanol (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.