Learn More
Sigma Organic Chemistry 3 5-Dimethylphenoxy ace 1G
Supplier: Sigma Organic Chemistry OTV0001241G

3 5-Dimethylphenoxy ace , 1G
About this item:
MDL #: MFCD09906402
Chemical Formula: C10H11NO
Molecular Weight: 161.2
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.