Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dimethylphenylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. AC431940500
Change view
Click to view available options
Quantity:
50 mL
Packaging:
AcroSeal™ Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC431940500 50 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC431940500 Supplier Thermo Scientific Chemicals Supplier No. 431940500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 34696-73-6 | C8H9BrMg | 209.37 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34696-73-6 , 109-99-9
Molecular Formula C8H9BrMg
Molecular Weight (g/mol) 209.37
MDL Number MFCD01311497
InChI Key XPXQQJMQESWIKM-UHFFFAOYSA-M
Synonym 3,5-dimethylphenylmagnesium bromide, xpxqqjmqeswikm-uhfffaoysa-m, 3,5-dimethylphenyl magnesium bromide, 3,5-dimethylphenylmagnesium bromide solution, 3,5-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 3,5-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,5-dimethylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
PubChem CID 10965748
IUPAC Name magnesium;1,3-dimethylbenzene-5-ide;bromide
SMILES CC1=CC([Mg]Br)=CC(C)=C1

Specifications

CAS Min % 9.2, 86.2
CAS Max % 13.8, 90.8
Color Yellow to Brown
Density 0.9100g/mL
Flash Point −18°C
Packaging AcroSeal™ Glass Bottle
Linear Formula (CH3)2C6H3MgBr
Quantity 50 mL
Specific Gravity 0.91
Solubility Information Solubility in water: reacts
Formula Weight 209.37
Physical Form Liquid
Chemical Name or Material 3, 5-Dimethylphenylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.
May form explosive peroxides.
Reacts violently with water.
Suspected of causing cancer.<br/
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wear protective gloves/protective clothing/eye p

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.