Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dinitrobenzoic acid, 98+%

Catalog No. AC153190010
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC153190010 1 kg Plastic bottle
AC153191000 100 g Plastic bottle
AC153195000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC153190010 Supplier Thermo Scientific Chemicals Supplier No. 153190010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 99-34-3 | C7H4N2O6 | 212.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-34-3
Molecular Formula C7H4N2O6
Molecular Weight (g/mol) 212.12
MDL Number MFCD00007253
InChI Key VYWYYJYRVSBHJQ-UHFFFAOYSA-N
Synonym 3,5-dinitrobenzoic acid, dinitrobenzoic acid, dnba, benzoic acid, 3,5-dinitro, 3-carboxy-1,5-dinitrobenzene, 3,5-dinitro-benzoic acid, unii-4v3f9q018p, ccris 3129, 3,5-dnba, 3,5-dinitrobenzenecarboxylic acid
PubChem CID 7433
ChEBI CHEBI:73914
IUPAC Name 3,5-dinitrobenzoic acid
SMILES OC(=O)C1=CC(=CC(=C1)[N+]([O-])=O)[N+]([O-])=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 205.0°C to 209.0°C
Density 1.6830g/mL
Assay Percent Range 98% min. (HPLC)
Packaging Plastic bottle
Linear Formula (O2N)2C6H3CO2H
Quantity 1 kg
Beilstein 09,413
Specific Gravity 1.683
Merck Index 15,3302
Solubility Information Solubility in water: 1350mg/l (25°C). Other solubilities: very soluble in alcohol and glacial acetic acid,sparingly soluble in ether,benzene and,carbon disulfide
Formula Weight 212.12
Percent Purity 98+%
Chemical Name or Material 3, 5-Dinitrobenzoic acid, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause long lasting harmful effects to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 202-751-1
RTECSNumber DG9140700
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.