Learn More
Sigma Organic Chemistry 3 6-Dibromo-1H-pyrazolo | 1G | MFCD22380424
Supplier: Sigma Organic Chemistry BLN000551G

3 6-Dibromo-1H-pyrazolo , 1G
About this Item:
MDL #: MFCD22380424
Molecular Weight: 276.92
UNSPSC Code: 12352200
Chemical Formula: C6H3Br2N3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.