Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole 95.0+%, TCI America™
SDP

Catalog No. D53421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D53421G 1 g
D5342200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D53421G Supplier TCI America Supplier No. D53421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 946491-48-1
Molecular Formula C26H27Br2NO
Molecular Weight (g/mol) 529.32
MDL Number MFCD30721573
InChI Key ACUVALSRGHGNLZ-UHFFFAOYNA-N
PubChem CID 68106047
IUPAC Name 3,6-dibromo-9-{4-[(2-ethylhexyl)oxy]phenyl}-9H-carbazole
SMILES CCCCC(CC)COC1=CC=C(C=C1)N1C2=C(C=C(Br)C=C2)C2=C1C=CC(Br)=C2

Specifications

Melting Point 59°C
Quantity 1 g
Formula Weight 529.32
Percent Purity ≥95.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Dibromo-9-[4-(2-ethylhexyloxy)phenyl]-9H-carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.