Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran

Catalog No. AC429610010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC429610010 1 g Glass bottle
AC429610050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC429610010 Supplier Thermo Scientific Chemicals Supplier No. 429610010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 862129-81-5 | C11H19BO2S | 226.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 862129-81-5
Molecular Formula C11H19BO2S
Molecular Weight (g/mol) 226.14
MDL Number MFCD10700154
InChI Key QZVRTORVCKGPPY-UHFFFAOYSA-N
Synonym 2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-dihydro-2h-thiopyran-4-ylboronic acid pinacol ester, 2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane, 2h-thiopyran, 3,6-dihydro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 6-dihydro-2h-thiopyran-4-yl-4, 3,6-dihydrothiopyran-4-boronic acid pinacol ester, 3,6-dihydro-2h-thiopyran-4-ylboronicacidpinacolester, 3,6-dihydro-2h-thiopyran-4-yl boronic acid pinacol ester, 2-3,6-dihydro-2h-thiopyran-4-yl-4,4,5,5,-tetramethyl-1,3,2-dioxaborolane
PubChem CID 46739735
IUPAC Name 2-(3,6-dihydro-2H-thiopyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(OC1(C)C)C1=CCSCC1

Specifications

Melting Point 42.0°C to 44.5°C
Packaging Glass bottle
Quantity 1 g
Formula Weight 226.15
Chemical Name or Material 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-thiopyran
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.