Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,6-Diphenylcarbazole 99.0+%, TCI America™
SDP

Catalog No. D44331G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D44331G 1 g
D44335G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D44331G Supplier TCI America Supplier No. D44331G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56525-79-2
Molecular Formula C24H17N
Molecular Weight (g/mol) 319.41
MDL Number MFCD00222619
InChI Key PCMKGEAHIZDRFL-UHFFFAOYSA-N
PubChem CID 3123295
IUPAC Name 3,6-diphenyl-9H-carbazole
SMILES N1C2=C(C=C(C=C2)C2=CC=CC=C2)C2=C1C=CC(=C2)C1=CC=CC=C1

Specifications

Melting Point 184°C
Color White
Quantity 1 g
Formula Weight 319.41
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3,6-Diphenylcarbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.