Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-[(6-Methylpyrazin-2-yl)oxy]benzylamine, 97%, Thermo Scientific™

Catalog No. CC62613CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC62613CB 250 mg Amber Glass Bottle
CC62613DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC62613CB Supplier Thermo Scientific Chemicals Supplier No. CC62613CB
Add to Cart

Chemical Identifiers

CAS 941716-89-8
Molecular Formula C12H13N3O
Molecular Weight (g/mol) 215.256
MDL Number MFCD09817526
InChI Key PJVRJIDLKXDROC-UHFFFAOYSA-N
Synonym 3-6-methylpyrazin-2-yl oxy benzylamine, 3-6-methylpyrazin-2-yl oxy phenyl methanamine, 1-3-6-methylpyrazin-2-yl oxy phenyl methanamine, 3-6-methylpyrazin-2-yloxy phenyl methylamine, 3-6-methylpyrazin-2-yl oxy phenyl methylamine
PubChem CID 24229681
IUPAC Name [3-(6-methylpyrazin-2-yl)oxyphenyl]methanamine
SMILES CC1=CN=CC(=N1)OC2=CC=CC(=C2)CN

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 215.25
Percent Purity 90%
Chemical Name or Material 3-[(6-Methylpyrazin-2-yl)oxy]benzylamine
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.