Learn More
US Plastic Corporation 3/8ID X 5/8OD TUBING 10FT
SDP
Supplier:  US Plastic Corporation 54037
Encompass
Natural, peroxide-cured siliconeWorking pressure @ 70°F: 10 psiTensile strength: 1444 psiElongation at break: 608%Durometer hardness (shore A): 50Temperature range: -100°F to 500°F (350°F max for NSF applications)Meets FDA standards & NSF 51 listedSold in increments of 10 feet. Shipped in full pieces
Catalog No. NC2332329
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.