Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,9-Diphenyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole 98.0+%, TCI America™
SDP

Catalog No. D50561G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50561G 1 g
D5056200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50561G Supplier TCI America Supplier No. D50561G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1359833-28-5
Molecular Formula C30H28BNO2
Molecular Weight (g/mol) 445.37
MDL Number MFCD29089360
InChI Key QVMVGEABPNCFOY-UHFFFAOYSA-N
Synonym 6,9-Diphenylcarbazole-3-boronic Acid Pinacol Ester, 4,4,5,5-Tetramethyl-2-(6,9-diphenylcarbazol-3-yl)-1,3,2-dioxaborolane
PubChem CID 71143518
IUPAC Name 3,9-diphenyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole
SMILES CC1(C)OB(OC1(C)C)C1=CC2=C(C=C1)N(C1=C2C=C(C=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164°C
Color White
Quantity 1 g
Formula Weight 445.37
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3,9-Diphenyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.