Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3'-Acetoxyacetophenone 98.0+%, TCI America™
SDP

Catalog No. A199625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A199625G 25 g
A19965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A199625G Supplier TCI America Supplier No. A199625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2454-35-5
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.187
MDL Number MFCD00017227
InChI Key OTHYPAMNTUGKDK-UHFFFAOYSA-N
PubChem CID 75563
IUPAC Name (3-acetylphenyl) acetate
SMILES CC(=O)C1=CC(=CC=C1)OC(=O)C

Specifications

Melting Point 46°C
Color White-Yellow
Boiling Point 147°C
Quantity 25 g
Formula Weight 178.19
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Acetoxyacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.