Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetyl-2,5-dichlorothiophene, 98%

Catalog No. AAA1386714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1386714 25 g
AAA1386706 5 g
AAA1386722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1386714 Supplier Thermo Scientific Chemicals Supplier No. A1386714
Add to Cart

CAS: 36157-40-1 | C6H4Cl2OS | 195.057 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36157-40-1
Molecular Formula C6H4Cl2OS
Molecular Weight (g/mol) 195.057
MDL Number MFCD00014522
InChI Key GYFDNIRENHZKGR-UHFFFAOYSA-N
Synonym 3-acetyl-2,5-dichlorothiophene, 1-2,5-dichlorothiophen-3-yl ethanone, 1-2,5-dichloro-3-thienyl ethan-1-one, 1-2,5-dichlorothiophen-3-yl ethan-1-one, 1-2,5-dichloro-3-thienyl ethanone, 2,5-dichloro-3-thienyl methyl ketone, ethanone, 1-2,5-dichloro-3-thienyl, 1-2,5-dichloro-thiophen-3-yl-ethanone, 1-2,5-dichlorothien-3-yl ethan-1-one, zlchem 481
PubChem CID 118920
IUPAC Name 1-(2,5-dichlorothiophen-3-yl)ethanone
SMILES CC(=O)C1=C(SC(=C1)Cl)Cl

Specifications

Melting Point 37°C to 39°C
Boiling Point 120°C to 122°C (4 mmHg)
Flash Point 110°C (230°F)
Quantity 25 g
Beilstein 121288
Formula Weight 195.07
Percent Purity 98%
Chemical Name or Material 3-Acetyl-2,5-dichlorothiophene
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 252-893-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.