Learn More
Medchemexpress LLC 3-acetyl-6-bromocoumarin | 2199-93-1 | MFCD00024075 | 98.0% | 267.08 g/mol | C11H7BrO3 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW1408695G
Encompass_Preferred
3-acetyl-6-bromocoumarin is a brominated coumarin research reagent supplied as a solid for biochemical and organic synthesis applications. It has reported antioxidant activity and shows inhibitory activity against human MAO-A in in vitro assays, with recommended storage at room temperature for the solid form.
  • Brominated coumarin scaffold for research applications.
  • Reported antioxidant activity (56%).
  • Inhibits human MAO-A in vitro (IC50 = 51.63 μM).
  • Solid, light yellow to yellow appearance.
  • Available in small-scale research quantities (for example, 5 G).
Catalog No. 50-002-83478