Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Acetylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. A194225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1942-25G 25 g
A19425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1942-25G Supplier TCI America Supplier No. A194225G
Add to Cart
Add to Cart

Specifications

CAS 204841-19-0
Melting Point 208°C
Color Yellow
Molecular Formula C8H9BO3
Quantity 25 g
MDL Number MFCD01074678
Synonym 3-acetylphenyl boronic acid, 3-acetylbenzeneboronic acid, 3-acetylphenylboronicacid, m-acetylphenylboronic acid, 3 '-boronoacetophenone
InChI Key SJGGDZCTGBKBCK-UHFFFAOYSA-N
SMILES B(C1=CC(=CC=C1)C(=O)C)(O)O
IUPAC Name (3-acetylphenyl)boronic acid
Molecular Weight (g/mol) 163.967
PubChem CID 2734310
ChEBI CHEBI:86552
Formula Weight 163.97
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.