Learn More
Sigma Organic Chemistry 3-Allyl-2-mercapto-5-4- 250MG
Supplier: Sigma Organic Chemistry OTV000550250MG

3-Allyl-2-mercapto-5-4- , 250mg
About this item:
MDL #: MFCD03964673
Chemical Formula: C16H14N2OS2
Molecular Weight: 314.43
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.