Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-1-propanol, 99%

Catalog No. p-7026808
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2304122 100 g
AAB2304136 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2304122 Supplier Thermo Scientific Chemicals Supplier No. B2304122
Only null left
Add to Cart

CAS: 156-87-6 | C3H9NO | 75.111 g/mol

3-Amino-1-propanol is used in the production of anionic emulsifiers, nonionic polyethylene emulsions. Its salts such as hydrochloride, nitrate, sulfate, phosphate are used for the balanced curing action with selected textile resins. It is used as a starting material in the preparation of beta-lactam antibiotics, humectants for foods and cosmetics. It acts as a corrosion inhibitor and finds application in water treatment, metal treatment and absorption of carbon dioxide gas.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-1-propanol is used in the production of anionic emulsifiers, nonionic polyethylene emulsions. Its salts such as hydrochloride, nitrate, sulfate, phosphate are used for the balanced curing action with selected textile resins. It is used as a starting material in the preparation of beta-lactam antibiotics, humectants for foods and cosmetics. It acts as a corrosion inhibitor and finds application in water treatment, metal treatment and absorption of carbon dioxide gas.

Solubility
Miscible with water, alcohol, ether, acetone and chloroform.

Notes
Hygroscopic. Incompatible withstrong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-87-6
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.111
MDL Number MFCD00008223
InChI Key WUGQZFFCHPXWKQ-UHFFFAOYSA-N
Synonym 3-aminopropanol, 3-amino-1-propanol, propanolamine, 1-propanol, 3-amino, 3-propanolamine, 1-amino-3-propanol, 3-hydroxypropylamine, 3-aminopropyl alcohol, 1,3-propanolamine, beta-alaninol
PubChem CID 9086
IUPAC Name 3-aminopropan-1-ol
SMILES C(CN)CO

Specifications

Melting Point 11°C
Density 0.983
Boiling Point 184°C to 187°C
Flash Point 101°C (214°F)
Odor Amine-like
Refractive Index 1.461
Quantity 100 g
UN Number UN2735
Beilstein 741855
Sensitivity Hygroscopic
Solubility Information Miscible with water,alcohol,ether,acetone and chloroform.
Formula Weight 75.11
Percent Purity 99%
Chemical Name or Material 3-Amino-1-propanol
Show More Show Less
Hazard Category H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H314-H318-H302-H312
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 205-864-4
RTECSNumber UA5600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.