Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2,6-dichloropyridine 98.0+%, TCI America™
SDP

Catalog No. A158025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A158025G 25 g
A15805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A158025G Supplier TCI America Supplier No. A158025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 62476-56-6
Molecular Formula C5H4Cl2N2
Molecular Weight (g/mol) 163.00
MDL Number MFCD00023417
InChI Key MJVZSRZTBDMYLX-UHFFFAOYSA-N
Synonym 3-amino-2,6-dichloropyridine, 2,6-dichloro-3-aminopyridine, 2,6-dichloro-3-pyridylamine, 3-pyridinamine, 2,6-dichloro, 2,6-dichloro-3-pyridinamine, 2,6-dichloropyridin-3-ylamine, 2,6-dichloro-pyridin-3-ylamine, 2,6-dichlorpyridin-3-amin, pubchem10384, ksc495k2f
PubChem CID 112846
IUPAC Name 2,6-dichloropyridin-3-amine
SMILES NC1=CC=C(Cl)N=C1Cl

Specifications

Melting Point 120°C
Quantity 25 g
Formula Weight 163.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2,6-dichloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.