Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-bromo-5-chloropyridine 98.0+%, TCI America™
SDP

Catalog No. A297325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A297325G 25 g
A29735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A297325G Supplier TCI America Supplier No. A297325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 90902-83-3
Molecular Formula C5H4BrClN2
Molecular Weight (g/mol) 207.455
MDL Number MFCD07368976
InChI Key AZOXQBMBDRLSCQ-UHFFFAOYSA-N
PubChem CID 2769640
IUPAC Name 2-bromo-5-chloropyridin-3-amine
SMILES C1=C(C(=NC=C1Cl)Br)N

Specifications

Melting Point 144°C
Quantity 25 g
Formula Weight 207.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-bromo-5-chloropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.