Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 98.0+%, TCI America™
SDP

Catalog No. A29931G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29931G 1 g
A2993200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29931G Supplier TCI America Supplier No. A29931G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 893440-50-1
Molecular Formula C12H19BN2O3
Molecular Weight (g/mol) 250.11
MDL Number MFCD12923427
InChI Key KYYKGOURQXPERA-UHFFFAOYSA-N
Synonym 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-amine, 3-amino-2-methoxypyridine-5-boronic acid pinacol ester, 5-amino-6-methoxypyridine-3-boronic acid pinacol ester, 2-methoxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-3-ylamine, 2-methoxy-3-aminopyridine-5-boronic acid pinacol ester, 5-amino-6-methoxypyridin-3-yl boronic acid pinacol ester, 2-methoxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-3-amine, 5-amino-6-methoxypyridine-3-boronic acid pinacol ester?, 3-amino-2-methoxypyridin-5-ylboronicacidpinacolester, 5-amino-6-methoxypyridine-3-boronicacidpinacolester
PubChem CID 59557920
IUPAC Name 2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-3-amine
SMILES COC1=C(N)C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 144°C
Quantity 1 g
Formula Weight 250.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-methoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.