Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-(3-bromophenyl)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. A25351G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A25351G 1 g
A25355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A25351G Supplier TCI America Supplier No. A25351G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 117391-50-1
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD00995843
InChI Key RLYAXKJHJUXZOT-UHFFFAOYSA-N
PubChem CID 603619
IUPAC Name 3-amino-3-(3-bromophenyl)propanoic acid
SMILES C1=CC(=CC(=C1)Br)C(CC(=O)O)N

Specifications

Melting Point 226°C
Color White
Quantity 1 g
Formula Weight 244.09
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-(3-bromophenyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.