Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-methyloxetane 97.0+%, TCI America™
SDP

Catalog No. A27921G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27921G 1 g
A2792200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A27921G Supplier TCI America Supplier No. A27921G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 874473-14-0
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.122
MDL Number MFCD11111713
InChI Key NQVWMPOQWBDSAI-UHFFFAOYSA-N
Synonym 3-methyl-3-oxetanamine, 3-amino-3-methyloxetane, 3-oxetanamine,3-methyl, 3-oxetanamine, 3-methyl, pubchem14416, pubchem23160, acmc-209qnl, 3-methyl 3-oxetanamine, 3-methyl-3-oxetaneamine, 3-amino-3-methyl-oxetane
PubChem CID 46835725
IUPAC Name 3-methyloxetan-3-amine
SMILES CC1(COC1)N

Specifications

Color Yellow
Quantity 1 g
Formula Weight 87.12
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Amino-3-methyloxetane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.