Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-3-(p-tolyl)propionic Acid 98.0+%, TCI America™
SDP

Catalog No. A234525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A234525G 25 g
A23455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A234525G Supplier TCI America Supplier No. A234525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 68208-18-4
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD01863249
InChI Key XPDAKEOBPKFUAH-SECBINFHSA-N
PubChem CID 2733680
IUPAC Name (3R)-3-azaniumyl-3-(4-methylphenyl)propanoate
SMILES CC1=CC=C(C=C1)[C@H]([NH3+])CC([O-])=O

Specifications

Melting Point 222°C
Color White
Quantity 25 g
Formula Weight 179.22
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-(p-tolyl)propionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.