Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-bromo-5-methylpyrazole 98.0+%, TCI America™
SDP

Catalog No. A29701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29701G 1 g
A29705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A29701G Supplier TCI America Supplier No. A29701G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1780-72-9
Molecular Formula C4H6BrN3
Molecular Weight (g/mol) 176.017
MDL Number MFCD03412169
InChI Key WZMBDMWFVPKYCF-UHFFFAOYSA-N
PubChem CID 1241299
IUPAC Name 4-bromo-5-methyl-1H-pyrazol-3-amine
SMILES CC1=C(C(=NN1)N)Br

Specifications

Melting Point 120°C
Quantity 1 g
Formula Weight 176.02
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-bromo-5-methylpyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.