Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-bromobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A244125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A244125G 25 g
A24415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A244125G Supplier TCI America Supplier No. A244125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2840-29-1
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.034
MDL Number MFCD00579106
InChI Key TZFJADFQEBASQU-UHFFFAOYSA-N
Synonym 3-amino-4-bromo-benzoic acid, benzoic acid, 3-amino-4-bromo, 3-amino-4-bromobenzoicacid, acmc-1cpid, 4-brom-3-aminobenzoesaure, ksc494k6h, 3-amino-4-bromobenzoic acid, 3-amino-4-bromo benzoic acid
PubChem CID 3418752
IUPAC Name 3-amino-4-bromobenzoic acid
SMILES C1=CC(=C(C=C1C(=O)O)N)Br

Specifications

Melting Point 228°C
Quantity 25 g
Formula Weight 216.03
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-bromobenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.