Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-chloro-N-(2-cyanoethyl)benzenesulfonamide 98%, Thermo Scientific™

Catalog No. AC163470050
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC163470050 5 g
AC163470100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC163470050 Supplier Thermo Scientific Chemicals Supplier No. 163470050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64415-13-0
Molecular Formula C9H10ClN3O2S
Molecular Weight (g/mol) 259.708
InChI Key DNMYCTLTLUHRRT-UHFFFAOYSA-N
PubChem CID 97449
IUPAC Name 3-amino-4-chloro-N-(2-cyanoethyl)benzenesulfonamide
SMILES C1=CC(=C(C=C1S(=O)(=O)NCCC#N)N)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Linear Formula H2NC6H3(Cl)SO2NHCH2CH2CN
Quantity 5 g
Synonym 3-amino-4-chloro-n-2-cyanoethyl benzenesulfonamide, 3-amino-4-chloro-n-2-cyanoethyl benzenesulphonamide, benzenesulfonamide, 3-amino-4-chloro-n-2-cyanoethyl, s-3-amino-4-chlorophenyl-2-cyanoethane-1-sulfonamido, 3-3-amino-4-chlorophenyl sulfonyl amino propanenitrile, benzenesulfonamide,3-amino-4-chloro-n-2-cyanoethyl, s-3-amino-4-chlorophenyl-2-cyanoethanesulfonamido
Formula Weight 259.7
Chemical Name or Material 3-Amino-4-chloro-N-(2-cyanoethyl)benzenesulfonamide, 98%
EINECSNumber 264-879-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.