Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-fluorobenzotrifluoride 98.0+%, TCI America™
SDP

Catalog No. A158225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A158225G 25 g
A15825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A158225G Supplier TCI America Supplier No. A158225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 535-52-4
Molecular Formula C7H5F4N
Molecular Weight (g/mol) 179.118
MDL Number MFCD00007653
InChI Key DRKWGMXFFCPZLW-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl aniline, 3-amino-4-fluorobenzotrifluoride, alpha,alpha,alpha,6-tetrafluoro-m-toluidine, 4-fluoro-3-aminobenzotrifluoride, 2-fluoro-5-trifluoromethyl benzenamine, 2-fluoro-5-trifluoromethyl phenylamine, benzenamine, 2-fluoro-5-trifluoromethyl, .alpha.,.alpha.,.alpha.,6-tetrafluoro-m-toluidine, pubchem1538, ksc494s7l
PubChem CID 136349
IUPAC Name 2-fluoro-5-(trifluoromethyl)aniline
SMILES C1=CC(=C(C=C1C(F)(F)F)N)F

Specifications

Boiling Point 190°C
Quantity 25 g
Formula Weight 179.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Amino-4-fluorobenzotrifluoride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.