Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-4-methylbenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. A0920250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0920250G 250 g
A092025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A0920250G Supplier TCI America Supplier No. A0920250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2458-12-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007742
InChI Key XKFIFYROMAAUDL-UHFFFAOYSA-N
Synonym 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh
PubChem CID 75568
IUPAC Name 3-amino-4-methylbenzoic acid
SMILES CC1=C(C=C(C=C1)C(=O)O)N

Specifications

Melting Point 166°C
Quantity 250 g
Formula Weight 151.17
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methylbenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.