Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-mercapto-1,2,4-triazole 98.0+%, TCI America™
SDP

Catalog No. A145525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A145525G 25 g
A14555G 5 g
A1455250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A145525G Supplier TCI America Supplier No. A145525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16691-43-3
Molecular Formula C2H4N4S
Molecular Weight (g/mol) 116.14
MDL Number MFCD00005231
InChI Key WZUUZPAYWFIBDF-UHFFFAOYSA-N
Synonym 3-amino-5-mercapto-1,2,4-triazole, iminothiourazole, 3-imino-5-thiourazole, 3-amino-1h-1,2,4-triazole-5-thiol, 5-amino-4h-1,2,4-triazole-3-thiol, 3-amino-1,2,4-triazole-5-thiol, s-triazole-3-thiol, 5-amino, 3h-1,2,4-triazole-3-thione, 5-amino-1,2-dihydro, 5-amino-1,2-dihydro-3h-1,2,4-triazole-3-thione, unii-353f2d8l1z
PubChem CID 2723869
IUPAC Name 5-amino-2,3-dihydro-1H-1,2,4-triazole-3-thione
SMILES NC1=NC(=S)NN1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 116.14
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-mercapto-1,2,4-triazole
EINECSNumber (5)-0607
RTECSNumber XZ5357500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.