Learn More
3-Amino-6-bromopyrazine-2-carboxylic acid, 98%, Thermo Scientific Chemicals
Description
3-Amino-6-bromopyrazine-2-carboxylic acid used to prepare 3-bromo-5-phenylpyrazine-2-amine. It serves as an intermediate in organic synthesis and pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3-Amino-6-bromopyrazine-2-carboxylic acid used to prepare 3-bromo-5-phenylpyrazine-2-amine. It serves as an intermediate in organic synthesis and pharmaceuticals.
Solubility
Soluble in dimethyl sulfoxide and methanol.
Notes
Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 178°C to 183°C |
| Color | Yellow |
| Quantity | 1 g |
| Beilstein | 147851 |
| Sensitivity | Light sensitive |
| Solubility Information | Soluble in dimethyl sulfoxide and methanol. |
| Formula Weight | 218.01 |
| Percent Purity | 98% |
| Physical Form | Solid |
| Chemical Name or Material | 3-Amino-6-bromopyrazine-2-carboxylic acid |
Safety and Handling
| Hazard Category | H302-H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c |
| TSCA | No |
| storageNote1 | -30° to -10°C |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.