Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-o-cresol 98.0+%, TCI America™
SDP

Catalog No. A182425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A182425G 25 g
A18245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A182425G Supplier TCI America Supplier No. A182425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53222-92-7
Molecular Formula C7H9NO
Molecular Weight (g/mol) 123.155
InChI Key FLROJJGKUKLCAE-UHFFFAOYSA-N
Synonym 3-amino-o-cresol, phenol, 3-amino-2-methyl, 2-amino-o-cresol, 2-methyl-3-aminophenol, 3-hydroxy-2-methylaniline, aminokresol, o-cresol, 3-amino, pubchem15039, 2-methyl-3-amino-phenol, 3-amino-2-methyl phenol
PubChem CID 104448
IUPAC Name 3-amino-2-methylphenol
SMILES CC1=C(C=CC=C1O)N

Specifications

Melting Point 133°C
Quantity 25 g
Formula Weight 123.16
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-o-cresol
EINECSNumber (3)-0707
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.