Learn More
Abcam 3-Aminobenzamide, PARP Inhibitor, >99% Purity, Solid, 100 mg
Description
- Purity: >99%
- Molecular Weight: 136.15 Da
- Biological Action: Poly(ADP-ribose) polymerase (PARP) inhibitor (Ki = 1.8 μM)
- Additional Effects: Stimulation of angiogenesis, neuroprotective and anti-necrotic effects, and free-radical scavenging ability
- Form: Solid
- CAS Number: 3544-24-9
- Storage: Store at ambient
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Poly(ADP-Ribose) Polymerase |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Content And Storage | Store at ambient temperature. |
| Buffer | 100% 3-Aminobenzamide |
| Form | Solid |
| Formulation | 100% 3-Aminobenzamide |
| Quantity | 100 mg |
| For Use With (Application) | PARP inhibition, angiogenesis stimulation, neuroprotection, anti-necrosis, free-radical scavenging |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.