Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobenzenethiol 97.0+%, TCI America™
SDP

Catalog No. A185225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A185225G 25 g
A18525G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A185225G Supplier TCI America Supplier No. A185225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22948-02-3
Molecular Formula C6H6NS
Molecular Weight (g/mol) 124.18
MDL Number MFCD00007791
InChI Key KFFUEVDMVNIOHA-UHFFFAOYSA-M
Synonym 3-aminothiophenol, benzenethiol, 3-amino, 3-mercaptoaniline, m-amino thiophenol, 3-aminobenzene-1-thiol, 3-amino thiophenol, 3-amino-benzenethiol, pubchem2470, acmc-1co9c
PubChem CID 31577
IUPAC Name (3-aminophenyl)sulfanide
SMILES NC1=CC=CC([S-])=C1

Specifications

Boiling Point 190°C
Quantity 25 g
UN Number 3267
Formula Weight 125.19
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-Aminobenzenethiol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.