Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminobiphenyl 99.0+%, TCI America™
SDP

Catalog No. B09871G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B09871G 1 g
B09875G 5 g
B098725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B09871G Supplier TCI America Supplier No. B09871G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2243-47-2
Molecular Formula C12H11N
Molecular Weight (g/mol) 169.23
MDL Number MFCD00047846
InChI Key MUNOBADFTHUUFG-UHFFFAOYSA-N
Synonym 3-Aminodiphenyl, 3-Biphenylamine
PubChem CID 16717
IUPAC Name [1,1'-biphenyl]-3-amine
SMILES NC1=CC=CC(=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Boiling Point 254°C
Quantity 1 g
Formula Weight 169.23
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Aminobiphenyl
RTECSNumber DU8900000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.