Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminophenylboronic Acid Monohydrate (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. A128125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1281-25G 25 g
A1281-5G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A1281-25G Supplier TCI America Supplier No. A128125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 206658-89-1
Molecular Formula C6H10BNO3
Molecular Weight (g/mol) 154.96
InChI Key XAEOVQODHLLNKX-UHFFFAOYSA-N
Synonym 3-aminophenylboronic acid monohydrate, 3-aminophenyl boronic acid hydrate, 3-aminophenylboronic acid hydrate, 3-aminobenzeneboronic acid monohydrate, m-aminophenylboronic acid hydrate
PubChem CID 14389423

Specifications

Quantity 25 g
MDL Number MFCD00007755
SMILES B(C1=CC(=CC=C1)N)(O)O.O
IUPAC Name (3-aminophenyl)boronic acid;hydrate
Formula Weight 136.95
Physical Form Crystalline Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.