Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopropyl Silica Gel (0.6-1.3mmol/g), TCI America™
SDP

Catalog No. A214025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A214025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A214025G Supplier TCI America Supplier No. A214025G
Add to Cart
Add to Cart

Chemical Identifiers

MDL Number MFCD00132846

Specifications

Color White
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material 3-Aminopropyl Silica Gel (0.6-1.3mmol/g)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.