Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Aminopropyl-Triethoxysilane, MP Biomedicals

Catalog No. ICN15476690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15476690 500 mL
ICN15476680 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15476690 Supplier MP Biomedicals, Inc Supplier No. 0215476690
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Forms aminopropyl derivative of glass, an adsorbent for affinity chromatography

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 919-30-2
Molecular Formula C9H23NO3Si
Molecular Weight (g/mol) 221.37
MDL Number MFCD00008207,MFCD01324904
InChI Key WYTZZXDRDKSJID-UHFFFAOYSA-N
Synonym 3-aminopropyltriethoxysilane, 3-aminopropyl triethoxysilane, aptes, 3-triethoxysilyl propan-1-amine, 1-propanamine, 3-triethoxysilyl, silicone a-1100, silane 1100, 3-triethoxysilyl propylamine, propylamine, 3-triethoxysilyl, triethoxy 3-aminopropyl silane
PubChem CID 13521
IUPAC Name (3-aminopropyl)triethoxysilane
SMILES CCO[Si](CCCN)(OCC)OCC

Specifications

Quantity 500 mL
Formula Weight 221.4
Concentration or Composition (by Analyte or Components) 1mL = approx. 0.95g
Physical Form Liquid
Chemical Name or Material 3-Aminopropyl-Triethoxysilane
Recommended Storage 0 to 5°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.