CAS RN 919-30-2
3-Aminopropyltriethoxysilane, 99%, AcroSeal™
CAS: 919-30-2 Molecular Formula: C9H23NO3Si Molecular Weight (g/mol): 221.37 MDL Number: MFCD00008207,MFCD01324904 InChI Key: WYTZZXDRDKSJID-UHFFFAOYSA-N Synonym: 3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane PubChem CID: 13521 IUPAC Name: 3-triethoxysilylpropan-1-amine SMILES: CCO[Si](CCCN)(OCC)OCC
3-Aminopropyltriethoxysilane, 99%
CAS: 919-30-2 Molecular Formula: C9H23NO3Si Molecular Weight (g/mol): 221.37 MDL Number: MFCD00008207,MFCD01324904 InChI Key: WYTZZXDRDKSJID-UHFFFAOYSA-N Synonym: 3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane PubChem CID: 13521 IUPAC Name: 3-triethoxysilylpropan-1-amine SMILES: CCO[Si](CCCN)(OCC)OCC
3-Aminopropyltriethoxysilane 98.0+%, TCI America™
CAS: 919-30-2 Molecular Formula: C9H23NO3Si Molecular Weight (g/mol): 221.37 MDL Number: MFCD00008207,MFCD01324904 InChI Key: WYTZZXDRDKSJID-UHFFFAOYSA-N Synonym: 3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane PubChem CID: 13521 IUPAC Name: (3-aminopropyl)triethoxysilane SMILES: CCO[Si](CCCN)(OCC)OCC
3-Aminopropyl-Triethoxysilane, MP Biomedicals
CAS: 919-30-2 Molecular Formula: C9H23NO3Si Molecular Weight (g/mol): 221.37 MDL Number: MFCD00008207,MFCD01324904 InChI Key: WYTZZXDRDKSJID-UHFFFAOYSA-N Synonym: 3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane PubChem CID: 13521 IUPAC Name: (3-aminopropyl)triethoxysilane SMILES: CCO[Si](CCCN)(OCC)OCC
(3-Aminopropyl)Triethoxysilane, Sigma-Aldrich
CAS: 919-30-2
(3-Aminopropyl)triethoxysilane, 98%
CAS: 919-30-2 Molecular Formula: C9H23NO3Si Molecular Weight (g/mol): 221.37 MDL Number: MFCD00008207,MFCD01324904 InChI Key: WYTZZXDRDKSJID-UHFFFAOYSA-N Synonym: 3-aminopropyltriethoxysilane,3-aminopropyl triethoxysilane,aptes,3-triethoxysilyl propan-1-amine,1-propanamine, 3-triethoxysilyl,silicone a-1100,silane 1100,3-triethoxysilyl propylamine,propylamine, 3-triethoxysilyl,triethoxy 3-aminopropyl silane PubChem CID: 13521 IUPAC Name: 3-triethoxysilylpropan-1-amine SMILES: CCO[Si](CCCN)(OCC)OCC
Sigma Aldrich (3-Aminopropyl)triethoxysilane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich (3-Aminopropyl)triethoxysilane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Sigma Aldrich (3-Aminopropyl)triethoxysilane
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.