Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(3-Aminopropyl)trimethoxysilane, 97%

Catalog No. p-7081348
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1128422 100 g
AAA1128436 500 g
AAA112840E 2500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1128422 Supplier Thermo Scientific Chemicals Supplier No. A1128422
Only null left
Add to Cart

CAS: 13822-56-5 | C6H17NO3Si | 179.291 g/mol

(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(3-Aminopropyl)trimethoxysilane is used as a silane coupling agent on silver nanoparticles. It is also used in silanization processes. It is also used to prepare surface modified titanate nanotubes (TiNT).

Solubility
Miscible with water.

Notes
Moisture sensitive. Incompatible withstrong acids, strong oxidizing agents, water, bases, alcohols and peroxides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13822-56-5
Molecular Formula C6H17NO3Si
Molecular Weight (g/mol) 179.291
MDL Number MFCD00008206
InChI Key SJECZPVISLOESU-UHFFFAOYSA-N
Synonym 3-aminopropyltrimethoxysilane, 3-aminopropyl trimethoxysilane, 3-trimethoxysilyl-1-propanamine, 3-trimethoxysilyl propan-1-amine, 3-trimethoxysilyl propylamine, 1-propanamine, 3-trimethoxysilyl, silane sc 3900, n-trimethoxysilylpropyl amine, propylamine, 3-trimethoxysilyl, unii-7mry0c9sb6
PubChem CID 83756
IUPAC Name 3-trimethoxysilylpropan-1-amine
SMILES CO[Si](CCCN)(OC)OC

Specifications

Density 1.02
Boiling Point 194°C
Flash Point 90°C (194°F)
Refractive Index 1.424
Linear Formula H2N(CH2)3Si(OCH3)3
Quantity 100 g
UN Number UN2735
Beilstein 2038118
Sensitivity Air and moisture sensitive
Solubility Information Miscible with water.
Formula Weight 179.29
Percent Purity 97%
Chemical Name or Material (3-Aminopropyl)trimethoxysilane
Show More Show Less
Hazard Category H227-H315-H318
Hazard Statement GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P264b-P280-P302+P352-P305+P351+P338-P310-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 237-511-5
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.